Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Streptavidin Agarose Columns, 1 mL PROMO

Catalog No. PI20351PM
Click to view available options
:
Thermo Scientific™ Pierce™ Streptavidin Agarose Columns

Bind biotinylated proteins and other ligands with a wide selection of high-quality agarose and acrylamide beads and columns of recombinant streptavidin biotin-binding protein.

Pierce Streptavidin Agarose and UltraLink Resins supply immobilized streptavidin biotin-binding protein in different resin formats, binding capacities and package sizes to suit nearly any immunoprecipitation or affinity purification procedure involving biotinylated molecules. The products use a recombinant form of streptavidin with a mass of 53 kDa and a near-neutral pI. The resins are prepared by covalently coupling streptavidin to the porous agarose or UltraLink Biosupport using efficient and stable chemistries, resulting in Streptavidin Agarose and UltraLink Resins that are leach-resistant and stable at pH 2-11. The products are excellent choices for a variety of small- or large-scale affinity purification applications involving biotinylated macromolecules, including separation of biotinylated molecules from samples and immunoprecipitation of antigens using biotin-labeled antibodies.

Highlights:

  • Proven coupling chemistries – the methods used to prepare these resins are superior to typical cyanogen bromide (CNBr) immobilization, providing high functionality, low nonspecific binding and excellent leach-resistance

  • Versatile resins – Agarose (polysaccharide-based) and UltraLink™ Biosupport (acrylamide-based) resins provide two different options for particular experimental conditions and are compatible with gravity-flow, spin and FPLC methods

  • Available in multiple formats – choose from two resin types, regular or high capacity preparations, and small or large-scale package sizes (bulk sizes are also available)

  • Superior performance – High Capacity Resin has 2 to 3 times higher binding capacity than alternative supplier products, allowing use of small amounts of resin for experiments

Item-Specific Features:

  • Agarose Resins are crosslinked 6% beaded agarose
  • UltraLink Resins beaded acrylamide support (UltraLink Biosupport)
  • Plus and High Capacity Resins have higher loading (i.e. density) of immobilized streptavidin protein than the regular forms

Recommended for:

Purification of membrane antigens in conjunction with biotinylated monoclonal antibodies; Cell-surface labeling with biotinylation reagents, followed by precipitation with immobilized streptavidin; Purification of cell-surface glycoproteins using biotinylated Concanavalin A; Recovery of single-stranded DNA for dideoxy sequencing

Specifications

Capacity (Metric) 5 mL
Content And Storage Contents: 5 x 1 mL prepacked Columns with twist-off bottom tab and accessory pack containing five white tips. Storage: Upon receipt Store at 4°C. Do not freeze the resin. Product is shipped at ambient temperature.
Particle Size 45 to 165 μm
Product Line Pierce™
Quantity 5 Columns
Protein Form Recombinant
Target Biotin-Tagged Molecule
Type Agarose
Stationary Phase Streptavidin
Sufficient For Binding 15 to 28 μg Biotin Per Column (1 to 3 mg Biotinylated BSA Per Column)
Format Spin Column
Column Type Affinity, Agarose Resin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.