Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Streptavidin, Hydrazide-Activated

Catalog No. pi21120
Click to view available options
Quantity:
2 mg

Perform Western blot, ELISA or other biotin-based detection with purified and HRP-, AP-, FITC-, TRITC-, PE- or APC-conjugated or maleimide-/hydrazide-activated streptavidin.

Thermo Scientific™ Pierce Streptavidin and Conjugates include purified recombinant streptavidin biotin-binding protein, plus standard SA-HRP, SA-AP and fluorescent conjugates for biotin-based assays.

These purified and conjugated forms of recombinant streptavidin protein enable detection of biotinylated antibodies and other probes in a variety of standard assay methods, including Western blotting, ELISA, immunohistochemistry and fluorescence imaging. Purified streptavidin and the streptavidin-HRP conjugates (peroxidase-conjugated streptavidin) are supplied lyophilized in phosphate-based buffers for immediate reconstitution with water. Alkaline Phosphatase conjugates are supplied as a liquid in a Tris-based buffer.

Highlights:

  • Recombinant streptavidin – high-quality protein, 53kDa, pI equals 6.8 to 7.5

  • Pure protein and enzyme conjugates – choose pure, unconjugated protein for biotin blocking and controls; enzyme conjugates for substrate-based detection

  • Fluorophore conjugates – standard fluorescent forms (fluorescein, rhodamine, etc.) for traditional immunofluorescence techniques

  • Activated preparations – choose a maleimide-activated or hydrazide-activated form to prepare a custom streptavidin conjugate with sulfhydryl- or carbohydrate-containing molecule

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Streptavidin
Description Pierce Streptavidin, Hydrazide Activated
Quantity 2 mg
Content And Storage Storage: Upon receipt store at room temperature or 4°C. Product shipped at ambient temperature.
Chemical Reactivity Ketone, Aldehyde
Reactive Moiety Amine, Hydrazide
Solubility Soluble in Water
Form Solid
Product Line Pierce™
Product Type Streptavidin Conjugate
Label or Dye Streptavidin
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.