Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Sulfo-SANPAH, No-Weigh™ Format

Catalog No. PI22589
Change view
Click to view available options
Quantity:
50 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI22589 50 mg
PIA35395 10 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI22589 Supplier Thermo Scientific™ Supplier No. 22589
Add to Cart
Add to Cart

Thermo Scientific™ Pierce Sulfo-SANPAH is a long-arm (18.2 Å) crosslinker that contains an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide.

Highlights:

  • Optimal photolysis occurs at 320-350nm, which limits damage to biomolecules by irradiation
  • Water-soluble
  • Reactive groups: sulfo-NHS ester and nitrophenyl azide (Amax= 470nm)
  • Reactive toward: amino groups and various sites
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Nonselective
Cleavable By Thiols
Molecular Weight (g/mol) 492.4
PEGylated No
Spacer Arm Length 18.2 Å
Content And Storage Upon receipt store at -20°C protected from light and moisture.
Cell Permeability No
Shipping Condition Ambient
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Sulfo-NHS Ester, Aryl Azide
Spacer Medium (10 to 30 Å)
Form Powder
Quantity 50 mg
Solubility Water
Format Standard, Single-use
Water Soluble Yes
Product Type Crosslinker
Show More Show Less
Can you provide the shelf-life for Sulfo-SANPAH?

Sulfo-SANPAH is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

What are the reactive groups on Sulfo-SANPAH and which groups do they react with?

Sulfo-SANPAH is a heterobifunctional crosslinker that has a sulfo-NHS ester and a photoactivatable nitrophenyl azide group. The sulfo-NHS ester reacts with primary amines in pH 7-9 buffers to form stable amide bonds. When exposed to UV light, nitrophenyl azides form a nitrene group that can initiate addition reactions with double bonds, insertion into C-H and N-H sites, or subsequent ring expansion to react with a nucleophile (e.g., primary amines).

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.