Learn More
Thermo Scientific™ Pierce™ Trifluoroacetic Acid (TFA), Sequencing grade, 100 g PROMO
Description
For complex peptide separations, the key to success can be to vary selectivity. Varying mobile phase composition on the same column can change selectivity enough to resolve peptide that would otherwise overlap. The TFA concentration is usually specified as 0.1% for reverse-phase HPLC of peptides. For reproducible separations from run-to-run or from lab-to-lab, it is essential to make concentrations the same.
Highlights:
- Purity: >99.5% TFA and exceptional clarity allows sensitive, nondestructive peptide detection at low UV wavelengths
- High-performance packaging: Packaged under nitrogen in amber glass with protective TFE-lined fluorocarbon caps to ensure TFA integrity
- Choice of formats for convenience: 1 mL ampules can prepare 1 L of 0.1% v/v TFA solution for the mobile phase in reverse-phase chromatography in moments
- CAS: 76-05-1
Storage Information
- Store in original container protected from direct sunlight in a dry, cool and well-ventilated area
- Should be stored;from 20° to 25°C
Specifications
Specifications
| Name Note | Trifluoroacetic Acid (TFA) |
| CAS | 76-05-01 |
| Molecular Weight (g/mol) | 114.02 |
| Percent Purity | 99.5% (TFA) |
| Quantity | 100 g |
| Physical Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.