Learn More
Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution, extended mass range PROMO
Description
The Thermo Scientific Pierce Triple Quadrupole Calibration Solution, Extended Mass Range, is a mixture of 14 highly pure, ionizable components (mass range: 69 m/z to 2800m/z) designed for both positive and negative ionization calibration of Thermo Scientific Triple Stage Quadrupole instruments.
- Improved mass accuracy - extended mass range designed to improve mass accuracy when compared to existing calibration solutions
- Positive and negative mode calibration - single formulation with extended mass ranges for both ionization calibration modes
- Highly validated - high-quality, rigorously tested formulation with lot-specific certificates of analysis
- Ready to use - reference standard may be directly loaded into a syringe and injected into the instrument
- Stable - store at 2 to 8°C for 18 months
The Pierce Triple Quadrupole Calibration Solution is a ready-to-use formulation that has been optimized for both positive and negative ionization calibration of instruments. The extended range masses (69 m/z to 2800m/z) improve performance of the Thermo Scientific Quantiva™ and Endura™ Triple Quadrupole mass spectrometers. Additionally, the Triple Quadrupole Calibration Solution, Extended Mass Range, is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle. The Pierce Triple Quadrupole Calibration Solution, Extended Mass Range, formulation contains 14 compounds that are ionizable in both positive and negative modes for instrument calibration. The formulation has been specifically optimized for use with the TSQ Endura™ Series and TSQ Quantiva™ Series instruments. The calibration solution is also compatible with the TSQ Quantum™ Series#, TSQ Discovery™ Series#, TSQ Quantum Ultra™ Series#, TSQ Quantum Access™ Series#, and TSQ Vantage™ Series# instruments using the list of recommended masses. Positive mode (m/Z): 69.0447, 142.1590, 322.0481, 622.0289, 922.0098. Negative mode (m/Z): 59.0128, 112.9845, 301.9970, 601.9779, 1033.9870. For the calibration procedure use: TSQ Series Getting Started Guide (70111-97152)
Specifications
Specifications
| For Use With (Equipment) | Mass Spectrometer |
| Product Type | Calibration Solution |
| Product Line | Pierce™ |
| Content And Storage | 1 x 10 mL, Store in refrigerator 2°C to 8°C). |
| Starting Material | Peptides, Small Molecules |
| Final Product Type | Peptides, Small Molecules |
| Detection Method | Mass Spectrometry |
| Workflow Step | Calibration |
| Quantity | 10 mL |
| Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.