Learn More
Thermo Scientific™ Pierce™ Triple Quadrupole Calibration Solution PROMO
Description
The Pierce Triple Quadrupole Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the required routine calibration to maintain the robust performance of Thermo Scientific Mass Spectrometers. The Triple Quadrupole Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.
Highlights
- Strong peaks — mixture of three tyrosine isotopes in methanol/formic acid solution
- Ready-to-use — load the reference standard into a syringe and inject into the instrument
- High-purity — mass spectrometry grade reagents in a a non-leachable container
- Stable — store at 2° to 8°C for more than one year
Specifications
Specifications
For Use With (Equipment) | Mass Spectrometer |
Product Type | Calibration Solution |
Product Line | Pierce™ |
Content And Storage | Store between 2°C and 8°C. |
Starting Material | Small Molecules |
Final Product Type | Small Molecules |
Detection Method | Mass Spectrometry |
Workflow Step | Calibration |
Quantity | 10 mL |
Form | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.