Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Universal Nuclease for Cell Lysis, 100 kU PROMO

Catalog No. PI88702PM
Click to view available options
:
Thermo Scientific™ Pierce™ Universal Nuclease for Cell Lysis

Catalog No. PI88702PM

Supplier: Thermo Scientific™ 88702

Ideal for a wide variety of applications where complete digestion of nucleic acids is needed and can be used in conjunction with cell lysis buffers.

Pierce Universal Nuclease for Cell Lysis is a genetically engineered endonuclease from Serratia marcescens. The enzyme is produced and purified from E. coli and consists of two identical 30-kDa subunits with two critical disulfide bonds. This indiscriminate endonuclease degrades single-stranded, double-stranded, linear and circular DNA and RNA and is effective over a wide range of temperatures and pH. This enzyme has very high specific activity (100-fold greater than DNase I) and has increased thermal stability compared to other nucleases. Pierce Universal Nuclease is ≥99% pure enzyme, is free of any measurable protease activity and is supplied at a concentration of 250 U/μL. Pierce Universal Nuclease for Cell Lysis is identical in performance to Benzonase™ Nuclease by EMD Merck™.

Highlights:

  • Broad spectrum – degrades all forms of DNA and RNA
  • Highest-quality enzyme – nuclease is at least 99% pure, as tested by SDS-PAGE
  • Robust activity – 100-fold greater specific activity than DNase I
  • Versatile – can be used with a wide variety of cell lysis reagents

Recommended for:

Use with B-PER, Y-PER or other commercial or homebrew cell lysis reagents and/or mechanical disruption to reduce viscosity in protein extracts; Remove DNA and RNA from recombinant protein preparations prior to downstream processing

Storage Requirements

Store in a cool, dry, well-ventilated area, protected from direct sunlight

Specifications

Product Type Cell Lysis Enzyme
Content And Storage Store in a cool, dry, well-ventilated area, protected from direct sunlight.
Form Liquid
Product Line Pierce™
Reagent Type Enzyme for Cell Lysis
Enzyme Nuclease
Quantity 100 kU

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.