Learn More
Thermo Scientific™ Pierce™ Yeast Digest Standard PROMO
Description
The Thermo Scientific Pierce Yeast Digest Standard is a lyophilized yeast peptide mixture ideal for monitoring LC/MS system performance.
Pierce Yeast Digest Standard features include:
- Positive control sample—well-characterized eukaryotic proteome sample protein digest (>3,000 proteins) for LC/MS method standardization, development, and troubleshooting
- Verified peptide quality—digestion conditions optimized for minimal missed cleavages or peptide modifications during processing
- Rigorously tested—high-quality, consistent protein digest guaranteed and documented via lot-specific certificates of analysis
- Convenient—ready-to-use lyophilized format
The Pierce Yeast Digest Standard is specifically formulated for LC/MS experiments and does not contain salts or detergents. The protein digest is derived from Saccharomyces cerevisiae yeast strain BY4741 (genetic background: MATa his3delta1 leu2delta0 met15delta0 ura3delta0). The yeast proteome is well-documented and characterized, and the digest is an ideal standard for more complex proteome mass spectrometry applications or when a non-mammalian digeste is required for analysis.
Using the standard routinely before analysis of complex samples makes it possible to monitor and normalize LC/MS performance between samples and over time.
Specifications
Specifications
| Chemical Name or Material | Yeast peptide digest |
| Packaging | Glass Vial |
| Physical Form | Lyophilized |
| Synonym | Yeast peptide digest |
| Quantity | 20 μg |
| Grade | MS |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.