Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIEZO1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP17837A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP17837A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP17837A70 Supplier Novus Biologicals Supplier No. NBP178537AF750
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIEZO1 Polyclonal specifically detects PIEZO1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PIEZO1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FAM38A, member A, membrane protein induced by beta-amyloid treatment, Mib, piezo-type mechanosensitive ion channel component 1
Gene Symbols PIEZO1
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human PIEZO1 protein (between residues 1300-1350) [UniProt Q92508]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9780.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.