Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIEZO1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP210504
Description
PIEZO1 Polyclonal specifically detects PIEZO1 in Human, Mouse, Rat, Porcine, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Knockdown Validated.Specifications
PIEZO1 | |
Polyclonal | |
Unconjugated | |
0.1M Tris 0.1M Glycine and 2% Sucrose with No Preservative | |
FAM38A, member A, membrane protein induced by beta-amyloid treatment, Mib, piezo-type mechanosensitive ion channel component 1 | |
Rabbit | |
Protein A purified | |
RUO | |
9780 | |
Lyophilized powder, reconstitute to 1 mg/ml in distilled water. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, Immunohistochemistry, KnockDown | |
LYOPH | |
Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:500, KnockDown Validated | |
Q92508 | |
PIEZO1 | |
Synthetic peptide derived from C-terminal domain of human Protein FAM38A. | |
0.1 mg | |
Primary | |
Reacts with human 287 kDa Protein FAM38A. Cross-reacts with mouse, rat, rabbit and other species due to sequence homology. Piezo-type mechanosensitive ion channel component 1. | |
Human, Mouse, Rat, Pig, Rabbit | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction