Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIEZO1 Recombinant Mouse Monoclonal Antibody (2-10-R-A)
GREENER_CHOICE

Catalog No. FEMA550642
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550642 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550642 Supplier Invitrogen™ Supplier No. MA550642
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Positive control: Recombinant protein lysates, rat brain tissue, mouse hippocampus tissue, mouse brain tissue. Predicted band size: 287 kDa Subcellular Location: Endoplasmic reticulum membrane Cell membrane.

Piezo1 is a pore-forming subunit of a mechanosensitive non-specific cation channel, and generates currents characterized by a linear current-voltage relationship that are sensitive to ruthenium red and gadolinium. Structurally, Piezo1 contains 36 transmembrane domains and functions as a homotetramer. Piezo1 plays a key role in epithelial cell adhesion by maintaining integrin activation through R-Ras recruitment to the ER, most probably in its activated state, and subsequent stimulation of calpain signaling. In the kidney, Piezo1 may contribute to the detection of intraluminal pressure changes and to urine flow sensing. Piezo1 acts as shear-stress sensor that promotes endothelial cell organization and alignment in the direction of blood flow through calpain activation and plays a key role in blood vessel formation and vascular structure in both development and adult physiology. The protein encoded by the Piezo1 gene is a mechanically-activated ion channel that links mechanical forces to biological signals. Defects in this gene have been associated with dehydrated hereditary stomatocytosis. Diseases associated with PIEZO1 include Dehydrated Hereditary Stomatocytosis with/without Pseudohyperkalemia And/Or Perinatal Edema and Lymphedema.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIEZO1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 2-10-R-A
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Piezo1
Gene Accession No. E2JF22, Q0KL00, Q92508
Gene Alias 9630020g22; DHS; Fam38a; family with sequence similarity 38, member A; KIAA0233; LMPH3; Membrane protein induced by beta-amyloid treatment; Mib; mKIAA0233; piezo type mechanosensitive ion channel component 1; Piezo1; piezo-type mechanosensitive ion channel component 1; Protein FAM38A; protein PIEZO1
Gene Symbols Piezo1
Host Species Mouse
Immunogen Recombinant protein within Human PIEZO1 aa 1275-1540.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234839, 361430, 9780
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.