Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIEZO2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2611300.025MG
Description
PIEZO2 Polyclonal specifically detects PIEZO2 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
PIEZO2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
PIEZO2 | |
Antibody was raised against a peptide corresponding to 19 amino acids near the amino terminus of human PIEZO2. Amino Acid Squence: HLTASLEKPEVRKLAEPGE | |
Affinity Purified | |
RUO | |
63895 | |
Human, Mouse | |
Purified |
Western Blot, ELISA | |
1 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
C18orf30, C18orf58, FAM38B, FAM38B2, family with sequence similarity 38, member B, family with sequence similarity 38, member B2, FLJ23144, FLJ23403, FLJ25916, FLJ34907, FLJ37734, FLJ45725, HsT748, HsT771, piezo-type mechanosensitive ion channel component 2, protein PIEZO2 | |
Rabbit | |
235 kDa | |
0.025 mg | |
Primary | |
PIEZO2 Antibody is predicted to not cross-react with PIEZO1. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction