Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA537136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA537136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA537136 Supplier Invitrogen™ Supplier No. PA537136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 70 kDa. Purity is >95% by SDS-PAGE.

PIF1 is a single-stranded (ss) DNA-dependent ATPase as well as a DNA helicase that unwinds DNA in a 5' to 3' direction with respect to the DNA strand on which it binds first. This protein is critical to DNA replication and telomere length maintenance in Saccharomyces cerevisiae. The PIF1 gene is highly conserved from yeast to humans. Mutations in the gene that encodes for PIF1 cause all telomeres to lengthen, suggesting that the PIF1 functions as a catalytic inhibitor of both new telomere formation and telomere elongation. Human PIF1 preferentially binds telomeric DNA where it inhibits telomerase activity. PIF1 specifically counteracts the RecQ homolog Sgs1 helicase activity, and degradation of PIF1 is mediated by the ubiquitin-26S proteasome pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIF1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene PIF1
Gene Accession No. Q1HG60, Q80SX8, Q9H611
Gene Alias 4631410M14; AI449441; ATP-dependent DNA helicase PIF1; ATP-dependent DNA helicase PIF1 {ECO:0000255; C15orf20; DNA helicase-like protein; DNA repair and recombination helicase PIF1; DNA repair and recombination helicase PIF1 {ECO:0000255; HAMAP-Rule:MF_03176}; petite integration frequency 1; PIF; Pif1; PIF1 5'-to-3' DNA helicase; PIF1 5'-to-3' DNA helicase homolog; PIF1 5'-to-3' DNA helicase homolog (S. cerevisiae); PIF1 homolog; Pif1/Rrm3 DNA helicase-like protein; Pif1/Rrm3 DNA-helicase-like protein
Gene Symbols PIF1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to residues in human PIF1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208084, 367645, 80119
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.