Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIF4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120777
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120777 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120777 Supplier Invitrogen™ Supplier No. PA5120777
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: seedling, rosette leaf. Immunogen sequence: IDYLKSLQLQ LQVMWMGSGM AAAAASAPMM FPGVQPQQFI RQIQSPVQLP RFPVMDQSAI QNNPGLVCQN PVQNQIISDR FARYIGGFPH MQAATQMQPM E.

Transcription factor acting negatively in the phytochrome B signaling pathway. May regulate the expression of a subset of genes involved in cell expansion by binding to the G-box motif. Activated by CRY1 and CRY2 in response to low blue light (LBL) by direct binding at chromatin on E-box variant 5'-CA[CT]GTG-3' to stimulate specific gene expression to adapt global physiology (e.g. hypocotyl elongation in low blue light). [Uniprot].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIF4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.