Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pig anti-Human IgG Fc Secondary Antibody, Novus Biologicals™
SDP

Pig Polyclonal Antibody

$281.84

Specifications

Dilution ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunoblot, Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Pig
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2-69484
SDP
View Documents
Novus Biologicals
NBP269484
10 mg
Each of 1 for $281.84
Only null left
Add to Cart
 
Description

Description

IgG Fc Polyclonal specifically detects IgG Fc in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunoblotting, Dot Blot.
Specifications

Specifications

ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoblotting 1:100 - 1:2000, Dot Blot 1:100-1:2000
Polyclonal
Pig
Secondary
ELISA, Immunocytochemistry, Immunofluorescence, Immunoblot, Dot Blot
Unconjugated
Human
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.