Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

New Pig Floor Sign and Marker
SDP

Catalog No. 19041549 Shop All New Pig Corporation Products
Change view
Click to view available options
Displayed Text:
Play It Smart Keep 6 ft. Apart
Please Wash Your Hands
Sanitize Hands Here
Single Direction Arrow, ONE WAY PLEASE
Take Precautions Thank You
Thank You for Covering Your Face
Thank You for Practicing Social Distancing
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Displayed Text
19-041-549 Please Wash Your Hands
19-041-552 Sanitize Hands Here
19-041-551 Play It Smart Keep 6 ft. Apart
19-041-547 Single Direction Arrow, ONE WAY PLEASE
19-041-548 Take Precautions Thank You
19-041-553 Thank You for Covering Your Face
19-041-550 Thank You for Practicing Social Distancing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-041-549 Supplier New Pig Supplier No. GMM21001BK

ADA Compliant Floor Signs and Markers are easy to install, stay down, and do not leave residue or damage floors. The low profile signs can be cleaned and will not create issues for heavy foot, cart, wheelchair, or walker traffic.

  • Helps companies and facilities to stay in compliance with social distancing guidelines
  • Creates a safe environment
  • Communicates to employees, guests & visitors how to safely navigate facilities
  • Withstands foot traffic, floor scrubbers, and disinfectant use
  • Will not become slippery when wet
  • Can be frequently cleaned with a scrubber, vacuum or broom without affecting performance
  • Sticks to most commercial floors including smooth indoor concrete, vct, linoleum, ceramic tile, quarry tile, and laminate flooring
  • Recommended for indoor & non-carpeted surfaces only
  • Lasts up to three months

Specifications

Material Polypropylene
Mounting Type Self-adhesive
For Use With (Application) To Remind and Reinforce Frequent Hand Washing
Shape Rectangular
Languages English
Dimensions (L x W) 17 x 24 in.
Displayed Text Please Wash Your Hands
Color White on Black
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.