Description
- Recycling system easily punctures aerosol cans, rendering them exempt from hazardous waste regulations when properly emptied
- Eliminates the risks associated with illegal aerosol disposal and the expense of hazardous waste
- Remove sediment and debris from stormwater before it enters your drains and inlets; ideal as a Best Management Practice (BMP) to help avoid violations
- Integrated magnetic strips on underside of filter allow easy installation in seconds; just set it in place — no lifting grate, no cable ties, no pinched fingers
- Natural coconut fibers are densely woven and vertically oriented to maximize
- Designed and tested to remove 99% of particulate size 0.425 mm and larger
- Built-in ports and scalloped edges help resist clogging; removable plugs offer overflow protection during high-flow storm events
- For curb-style inlets with grate, align magnets and place straight (unscalloped) edge against curb for best performance
- For curb-style inlets without a grate, filter can be folded in half inserted into opening; friction fit holds filter in place
- Reusable product can be washed and reinstalled for extended service
- Minimizes worker injuries caused by fire, explosion or VOC release from improperly emptied or unemptied cans
- System works with domed mini, standard and jumbo aerosol cans
- Combination Coalescing/Carbon Filter captures odors and potentially harmful VOCs released by punctured aerosol cans
- Timestrip indicator eliminates the guesswork of carbon cartridge changeouts
- Combination Coalescing/Carbon Filter life is 3 months
- Angled hardened steel pin punctures cans cleanly without leaving jagged edges
- Unit mounts directly into 2 in. bung on a 30- or 55-gallon closed-head steel drum; residual liquids drain directly into drum Included ground wire helps prevent static electricity buildup
- Selling scrap steel can help you meet waste minimization goals at your facility
Specifications
Specifications
| Type | Sediment Catcher |
| Weight (English) | 13 lb. |
| Weight (Metric) | 5.9 kg |