Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG3 Antibody (1C9), Alexa Fluor™ 594, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP222591AF594

Catalog No. NP22291A94


Only null left
Add to Cart

Description

Description

PIG3 Monoclonal specifically detects PIG3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

PIG3
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
TP53I3
Recombinant human TP53I3 (1-332aa) purified from E. coli
0.1 mL
Primary
Human
IgG2a κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
1C9
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
p53-induced gene 3 protein, PIG3quinone oxidoreductase homolog, quinone oxidoreductase PIG3, tumor protein p53 inducible protein 3, Tumor protein p53-inducible protein 3
Mouse
Protein G purified
Apoptosis
9540.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.