Learn More
Invitrogen™ PIG3 Monoclonal Antibody (OTI3B11)

Description
PIG3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PIG3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3B11 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TP53I3 |
| Gene Accession No. | Q53FA7 |
| Gene Alias | p53-induced gene 3 protein; PIG3; quinone oxidoreductase homolog; quinone oxidoreductase PIG3; Quinone oxidoreductase-like protein 2; RGD1304982; TP53I3; tumor protein p53 inducible protein 3; Tumor protein p53-inducible protein 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.