Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIG3 (p53 inducible gene 3) Rabbit anti-Human, Polyclonal, Exalpha

Rabbit Polyclonal Antibody
Supplier: Vector Laboratories X1155P

This item is not returnable.
View return policy
Description
PIG3 (p53 inducible gene 3) Polyclonal antibody specifically detects PIG3 (p53 inducible gene 3) in Human samples. It is validated for Western BlotSpecifications
PIG3 (p53 inducible gene 3) | |
Polyclonal | |
Unconjugated | |
TP53I3 | |
p53-induced gene 3 protein; PIG3; quinone oxidoreductase homolog; Quinone oxidoreductase PIG3; Quinone oxidoreductase-like protein 2; RGD1304982; TP53I3; tumor protein p53 inducible protein 3; Tumor protein p53-inducible protein 3 | |
Antibody developed using full-length recombinant PIG-3 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.5-2 mg/mL | |
PBS with 0.08% sodium azide; pH | |
Q53FA7 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
9540 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction