Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIGA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551527
Description
Immunogen sequence: FADVSSVLTN KLLTVSLCDT NHIICVSYTS KENTVLRAAL NPEIVSVIPN AVDPTDFTPD PFRRHDSITI VVVSRLVYRK GIDLLSGIIP ELCQKYPDLN FIIGGEGPKR II Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
This gene encodes a protein required for synthesis of N-acetylglucosaminyl phosphatidylinositol (GlcNAc-PI), the first intermediate in the biosynthetic pathway of GPI anchor. The GPI anchor is a glycolipid found on many blood cells and which serves to anchor proteins to the cell surface. Paroxysmal nocturnal hemoglobinuria, an acquired hematologic disorder, has been shown to result from mutations in this gene. Alternate splice variants have been characterized.
Specifications
PIGA | |
Polyclonal | |
Unconjugated | |
PIGA | |
AI194334; class A GlcNAc-inositol phospholipid assembly protein; GLCNAC-PI synthesis protein; GPI anchor biosynthesis; GPI3; MCAHS2; N-acetylglucosaminyl-phosphatidylinositol biosynthetic protein; phosphatidylinositol glycan anchor biosynthesis class A; phosphatidylinositol glycan anchor biosynthesis, class A; phosphatidylinositol glycan anchor biosynthesis, class A (paroxysmal nocturnal hemoglobinuria); phosphatidylinositol glycan, class A; phosphatidylinositol N-acetylglucosaminyltransferase subunit A; Phosphatidylinositol-glycan biosynthesis class A protein; phosphatidylinositol-glycan biosynthesis, class A protein; phosphatidylinositolglycan class A; PIGA; PIG-A; PNH1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5277 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P37287 | |
PIGA | |
Recombinant protein corresponding to Human PIGA. Recombinant protein control fragment (Product #RP-91907). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction