Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGA Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104006
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104006 Supplier Invitrogen™ Supplier No. PA5104006
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PIGA.

This gene encodes a protein required for synthesis of N-acetylglucosaminyl phosphatidylinositol (GlcNAc-PI), the first intermediate in the biosynthetic pathway of GPI anchor. The GPI anchor is a glycolipid found on many blood cells and which serves to anchor proteins to the cell surface. Paroxysmal nocturnal hemoglobinuria, an acquired hematologic disorder, has been shown to result from mutations in this gene. Alternate splice variants have been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PIGA
Gene Accession No. P37287, Q64323
Gene Alias AI194334; class A GlcNAc-inositol phospholipid assembly protein; GLCNAC-PI synthesis protein; GPI anchor biosynthesis; GPI3; MCAHS2; N-acetylglucosaminyl-phosphatidylinositol biosynthetic protein; phosphatidylinositol glycan anchor biosynthesis class A; phosphatidylinositol glycan anchor biosynthesis, class A; phosphatidylinositol glycan anchor biosynthesis, class A (paroxysmal nocturnal hemoglobinuria); phosphatidylinositol glycan, class A; phosphatidylinositol N-acetylglucosaminyltransferase subunit A; Phosphatidylinositol-glycan biosynthesis class A protein; phosphatidylinositol-glycan biosynthesis, class A protein; phosphatidylinositolglycan class A; PIGA; PIG-A; PNH1
Gene Symbols PIGA
Host Species Rabbit
Immunogen A synthesized peptide derived from human PIGA(Accession P37287), corresponding to amino acid residues T446-R484.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18700, 363464, 5277
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.