Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIGB Monoclonal Antibody (2G7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524388
Description
Peptide Sequence: TSFVPDEYWQ SLEVSHHMVF NYGYLTWEWT ERLRSYTYPL IFASIYKILH LLGKDSVQLL IWIPRLAQAL LSAVADVRLY SLMKQLENQE VAR.
Mannosyltransferase involved in glycosylphosphatidylinositol-anchor biosynthesis. Transfers the third alpha-1, 2-mannose to Man2-GlcN-acyl-PI during GPI precursor assembly.
Specifications
PIGB | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q92521 | |
PIGB | |
PIGB (NP_004846, 80 a.a. ~ 172 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2G7 | |
Unconjugated | |
PIGB | |
dol-P-Man dependent GPI mannosyltransferase; GPI mannosyltransferase 3; GPI mannosyltransferase III; GPI-MT-III; involved in GPI-anchor biosynthesis; phosphatidylinositol glycan anchor biosynthesis class B; phosphatidylinositol glycan anchor biosynthesis, class B; phosphatidylinositol glycan, class B; phosphatidylinositol-glycan biosynthesis class B protein; Pigb; Pig-b | |
Mouse | |
Protein A | |
RUO | |
9488 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction