Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGC Monoclonal Antibody (1G2)
GREENER_CHOICE

Catalog No. PIMA519600
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519600 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519600 Supplier Invitrogen™ Supplier No. MA519600
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MYAQPVTNTK EVKWQKVLYE RQPFPDNYVD RRFLEELRKN IHARKYQYWA VVFE.

This gene encodes an endoplasmic reticulum associated protein that is involved in glycosylphosphatidylinositol (GPI) lipid anchor biosynthesis. The GPI lipid anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. The encoded protein is one subunit of the GPI N-acetylglucosaminyl (GlcNAc) transferase that transfers GlcNAc to phosphatidylinositol (PI) on the cytoplasmic side of the endoplasmic reticulum. Two alternatively spliced transcripts that encode the same protein have been found for this gene. A pseudogene on chromosome 11 has also been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGC
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1G2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PIGC
Gene Accession No. Q92535
Gene Alias 2.4.1.198; 3110030E07Rik; AW212108; GPI2; phosphatidylinositol glycan anchor biosynthesis class C; phosphatidylinositol glycan anchor biosynthesis, class C; phosphatidylinositol glycan, class C; phosphatidylinositol N-acetylglucosaminyltransferase subunit C; Phosphatidylinositol-glycan biosynthesis class C protein; phosphatidylinositol-glycan biosynthesis, class C protein; PIGC; PIG-C
Gene Symbols PIGC
Host Species Mouse
Immunogen PIGC (NP_002633, 1 a.a. ~ 54 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5279
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.