Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568267 Supplier Invitrogen™ Supplier No. PA568267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

This gene encodes an endoplasmic reticulum associated protein that is involved in glycosylphosphatidylinositol (GPI) lipid anchor biosynthesis. The GPI lipid anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. The encoded protein is one subunit of the GPI N-acetylglucosaminyl (GlcNAc) transferase that transfers GlcNAc to phosphatidylinositol (PI) on the cytoplasmic side of the endoplasmic reticulum. Two alternatively spliced transcripts that encode the same protein have been found for this gene. A pseudogene on chromosome 11 has also been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGC
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PIGC
Gene Accession No. Q5PQQ4, Q92535, Q9CXR4
Gene Alias 2.4.1.198; 3110030E07Rik; AW212108; GPI2; phosphatidylinositol glycan anchor biosynthesis class C; phosphatidylinositol glycan anchor biosynthesis, class C; phosphatidylinositol glycan, class C; phosphatidylinositol N-acetylglucosaminyltransferase subunit C; Phosphatidylinositol-glycan biosynthesis class C protein; phosphatidylinositol-glycan biosynthesis, class C protein; PIGC; PIG-C
Gene Symbols PIGC
Host Species Rabbit
Immunogen Synthesized peptide derived from human PIGC.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364032, 5279, 67292
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.