Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGC Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572316
Description
This gene encodes an endoplasmic reticulum associated protein that is involved in glycosylphosphatidylinositol (GPI) lipid anchor biosynthesis. The GPI lipid anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. The encoded protein is one subunit of the GPI N-acetylglucosaminyl (GlcNAc) transferase that transfers GlcNAc to phosphatidylinositol (PI) on the cytoplasmic side of the endoplasmic reticulum. Two alternatively spliced transcripts that encode the same protein have been found for this gene. A pseudogene on chromosome 11 has also been characterized.
Specifications
PIGC | |
Polyclonal | |
Unconjugated | |
PIGC | |
Phosphatidylinositol N-acetylglucosaminyltransferase subunit C; 2.4.1.198; Phosphatidylinositol-glycan biosynthesis class C protein; PIG-C; PIGC; GPI2 | |
Rabbit | |
Protein A | |
RUO | |
5279 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Western Blot | |
0.75 mg/mL | |
PBS with 0.09% sodium azide | |
Q92535 | |
PIGC | |
KLH conjugated synthetic peptide between 264-294 amino acids from human PIGC | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction