Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGF Antibody (RM0010-8F09), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB11085468V
Description
PIGF Monoclonal specifically detects PIGF in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
PIGF | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PIGF | |
Mouse recombinant PlGF protein. | |
0.1 mL | |
Primary | |
This recognizes mouse PlGF. | |
Store at 4C in the dark. | |
IgG2 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
RM0010-8F09 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
GPI11 homolog, MGC32646, MGC33136, phosphatidylinositol glycan anchor biosynthesis, class F, phosphatidylinositol glycan, class F, phosphatidylinositol-glycan biosynthesis class F protein, PIG-F | |
Rat | |
Protein G purified | |
Biologically Active Proteins | |
5281 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction