Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569078 Supplier Invitrogen™ Supplier No. PA569078
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Horse: 92%; Human: 100%; Mouse: 86%; Pig: 86%; Rat: 86%; Sheep: 86%.

Phosphatidylinositol-glycans(PIGs) aremulti-passtransmembrane proteins that localize to the endoplasmicreticulum. PIGs exhibitvariousfunctions but all are crucial for the biosynthesis of the glycosylphosphatidylinositol (GPI) anchor. Some PIG proteins are components of the GPI transamidase complex and play a role in the recognition of either the GPI attachmentsignal or the lipid portion of GPI. Other PIGs belong to the glycosyltransferase complex and function in the transfer of N-acetylglucosamine (GlcNAc) to phosphatidyli nositol (PI). A variety of other PIGs play distinct rolesin GPIsynthesis. PIG-F functions as an auxiliary subunit of ethanolamine phosphate (EtNP) transferases. It associates with PIG-O and isrequired for its expression and stability. Together these two PIGs function as an EtNP transferase and catalyze the transfer of EtNP to the third mannose (Man-3) of GPI. Amutation in the gene encoding PIG-F may result in a block of EtNP addition to Man-3 and lead to an absence of GPI-anchored proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGF
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PIGF
Gene Accession No. Q07326
Gene Alias GPI11 homolog; phosphatidylinositol glycan anchor biosynthesis class F; phosphatidylinositol glycan anchor biosynthesis, class F; phosphatidylinositol glycan, class F; phosphatidylinositol-glycan biosynthesis class F protein; phosphatidylinositol-glycan biosynthesis class protein-like protein; PIGF; PIG-F; Unknown (protein for MGC:133893)
Gene Symbols PIGF
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human PIGF.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5281
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.