Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIGF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575899
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Phosphatidylinositol-glycans(PIGs) aremulti-passtransmembrane proteins that localize to the endoplasmicreticulum. PIGs exhibitvariousfunctions but all are crucial for the biosynthesis of the glycosylphosphatidylinositol (GPI) anchor. Some PIG proteins are components of the GPI transamidase complex and play a role in the recognition of either the GPI attachmentsignal or the lipid portion of GPI. Other PIGs belong to the glycosyltransferase complex and function in the transfer of N-acetylglucosamine (GlcNAc) to phosphatidyli nositol (PI). A variety of other PIGs play distinct rolesin GPIsynthesis. PIG-F functions as an auxiliary subunit of ethanolamine phosphate (EtNP) transferases. It associates with PIG-O and isrequired for its expression and stability. Together these two PIGs function as an EtNP transferase and catalyze the transfer of EtNP to the third mannose (Man-3) of GPI. Amutation in the gene encoding PIG-F may result in a block of EtNP addition to Man-3 and lead to an absence of GPI-anchored proteins.
Specifications
PIGF | |
Polyclonal | |
Unconjugated | |
PIGF | |
GPI11 homolog; phosphatidylinositol glycan anchor biosynthesis class F; phosphatidylinositol glycan anchor biosynthesis, class F; phosphatidylinositol glycan, class F; phosphatidylinositol-glycan biosynthesis class F protein; phosphatidylinositol-glycan biosynthesis class protein-like protein; PIGF; PIG-F; Unknown (protein for MGC:133893) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18701, 5281, 681086 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.3 | |
O09101, Q07326 | |
PIGF | |
Synthetic peptide corresponding to residues in Human PIGF. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction