Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGH Monoclonal Antibody (2F8)
PIGH Monoclonal Antibody (2F8)
Supplier: Thermo Scientific MA519601
Description

Specifications
PIGH | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q14442 | |
PIGH | |
PIGH (NP_004560, 89 a.a. ~ 188 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
2F8 | |
Unconjugated | |
PIGH | |
2210416H01Rik; A930028P05Rik; GPI-H; Mrp64; phosphatidylinositol glycan anchor biosynthesis class H; phosphatidylinositol glycan anchor biosynthesis, class H; phosphatidylinositol glycan, class H; phosphatidylinositol N-acetylglucosaminyltransferase subunit H; Phosphatidylinositol-glycan biosynthesis class H protein; phosphatidylinositol-glycan biosynthesis, class H protein; Pigh; PIG-H | |
Mouse | |
Affinity chromatography | |
RUO | |
5283 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction