Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556802 Supplier Invitrogen™ Supplier No. PA556802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KIDQETLLII DSLGIQMTSS YASGKESTTF IEMGKVKDIV INEAIYMQKV IYYLCILLKD PVEPHGISQV VPVFQSAKPR LDCLIEVYRS CQEILAHQKA TSTS Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

This gene encodes an endoplasmic reticulum associated protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI anchor is a glycolipid found on many blood cells and which serves to anchor proteins to the cell surface. The protein encoded by this gene is a subunit of the GPI N-acetylglucosaminyl (GlcNAc) transferase that transfers GlcNAc to phosphatidylinositol (PI) on the cytoplasmic side of the endoplasmic reticulum. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGH
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIGH
Gene Accession No. Q14442
Gene Alias 2210416H01Rik; A930028P05Rik; GPI-H; Mrp64; phosphatidylinositol glycan anchor biosynthesis class H; phosphatidylinositol glycan anchor biosynthesis, class H; phosphatidylinositol glycan, class H; phosphatidylinositol N-acetylglucosaminyltransferase subunit H; Phosphatidylinositol-glycan biosynthesis class H protein; phosphatidylinositol-glycan biosynthesis, class H protein; Pigh; PIG-H
Gene Symbols PIGH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIGH. Recombinant protein control fragment (Product #RP-89081).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5283
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.