Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIGK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111433
Description
Immunogen sequence: CVSTPGHRTD LFQRDPKNVL ITDFFGSVRK VEITTETIKL QQDSEIMESS YKEDQMDEKL MEPLKYAEQL PVAQIIHQKP KLKD.
This gene encodes a member of the cysteine protease family C13 that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is a member of the multisubunit enzyme, GPI transamidase and is thought to be its enzymatic component. GPI transamidase mediates GPI anchoring in the endoplasmic reticulum, by catalyzing the transfer of fully assembled GPI units to proteins.
Specifications
PIGK | |
Polyclonal | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92643 | |
PIGK | |
Recombinant Protein corresonding to Human PIGK. Recombinant protein control fragment (Product #RP-101796). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
PIGK | |
3000001O05Rik; GPI transamidase; GPI transamidase subunit; GPI8; GPI8 homolog; GPI-anchor transamidase; hGPI8; phosphatidylinositol glycan anchor biosynthesis class K; phosphatidylinositol glycan anchor biosynthesis, class K; phosphatidylinositol glycan, class K; phosphatidylinositol-glycan biosynthesis class K protein; Pigk; PIG-K | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10026 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction