Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552976 Supplier Invitrogen™ Supplier No. PA552976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKSREQGGRL GAESRTLLVI AHPDDEAMFF APTVLGLARL RHWVYLLCFS AGNYYNQGET RKKELLQSCD VLGIPLSSVM IIDNRDFPDD PGMQWDTEHV ARVLLQHIEV NGI Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 74%.

This gene encodes an enzyme that catalyzes the second step of glycosylphosphatidylinositol (GPI) biosynthesis, which is the de-N-acetylation of N-acetylglucosaminylphosphatidylinositol (GlcNAc-PI). Study of a similar rat enzyme suggests that this protein localizes to the endoplasmic reticulum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGL
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIGL
Gene Accession No. Q9Y2B2
Gene Alias CHIME; N-acetylglucosaminylphosphatidylinositol deacetylase; N-acetylglucosaminyl-phosphatidylinositol de-N-acetylase; phosphatidylinositol glycan anchor biosynthesis class L; phosphatidylinositol glycan anchor biosynthesis, class L; phosphatidylinositol glycan, class L; phosphatidylinositol-glycan biosynthesis class L protein; PIGL; PIG-L
Gene Symbols PIGL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIGL. Recombinant protein control fragment (Product #RP-90099).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9487
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.