Learn More
Invitrogen™ PIGM Polyclonal Antibody

Description
Positive test controls include: BxPC-3, Mouse thymus, Mouse spleen, Mouse lung, Rat thymus, Rat spleen. Immunogen sequence: YGVFQDRTLH VRYTDIDYQV FTDAARFVTE GRSPYLRATY RYTPLLGWLL TPNIYLSELF GK.
Specifications
Specifications
| Antigen | PIGM |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | PIGM |
| Gene Accession No. | Q8C2R7, Q9EQY6, Q9H3S5 |
| Gene Alias | 4933437L05Rik; AI644422; AV302080; C920011G20Rik; dol-P-Man dependent GPI mannosyltransferase; DPM:GlcN-(acyl-)PI mannosyltransferase; GPI mannosyltransferase 1; GPI mannosyltransferase I; GPI-MT-I; phosphatidylinositol glycan anchor biosynthesis class M; phosphatidylinositol glycan anchor biosynthesis, class M; phosphatidylinositol glycan, class M; phosphatidylinositol-glycan biosynthesis class M protein; Pigm; PIG-M; PIG-M mannosyltransferase; PIG-M mRNA for mannosyltransferase |
| Gene Symbols | PIGM |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.