Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIGN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559149
Description
Immunogen sequence: KVDDGVKEIV SMFNHFYGND GKTTFIFTSD HGMTDWGSHG AGHPSETLTP LVTWGAGIKY PQRVSAQQFD DAFLKEWRLE NWK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
PIGN is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is expressed in the endoplasmic reticulum and transfers phosphoethanolamine (EtNP) to the first mannose of the GPI anchor.
Specifications
PIGN | |
Polyclonal | |
Unconjugated | |
PIGN | |
Gm20308; GPI ethanolamine phosphate transferase 1; MCAHS; MCAHS1; MCD4; MCD4 homolog; MDC4; phosphatidylinositol glycan anchor biosynthesis class N; phosphatidylinositol glycan anchor biosynthesis, class N; phosphatidylinositol glycan, class N; phosphatidylinositol-glycan biosynthesis class N protein; PIGN; PIG-N | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23556 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95427 | |
PIGN | |
Recombinant protein corresponding to Human PIGN. Recombinant protein control fragment (Product #RP-98290). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction