Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGO Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116882
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116882 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116882 Supplier Invitrogen™ Supplier No. PA5116882
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa, 22Rv1, Mouse uterus, Mouse spleen Immunogen sequence: FAVGCPLLLL WPFLCESQGL RKRQQPPGNE ADARVRPEEE EEPLMEMRLR DAPQHFYAAL LQLGLKYLFI LGIQILACAL AASILRRHLM VWKVFAPKFI F.

This gene encodes a protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid which contains three mannose molecules in its core backbone. The GPI-anchor is found on many blood cells and serves to anchor proteins to the cell surface. This protein is involved in the transfer of ethanolaminephosphate (EtNP) to the third mannose in GPI. At least two alternatively spliced transcripts encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGO
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PIGO
Gene Accession No. Q8TEQ8, Q9JJI6
Gene Alias GPI ethanolamine phosphate transferase 3; HPMRS2; phosphatidylinositol glycan anchor biosynthesis class O; phosphatidylinositol glycan anchor biosynthesis, class O; phosphatidylinositol glycan, class O; phosphatidylinositol-glycan biosynthesis class O protein; Pigo; PIG-O; RP11-182N22.4; UNQ632/PRO1249
Gene Symbols PIGO
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 950-1050 of human PIGO (NP_1160232).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56703, 84720
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.