Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGO Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$148.20 - $378.00
Specifications
Antigen | PIGO |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
PIGO Polyclonal antibody specifically detects PIGO in Human, Mouse samples. It is validated for Western BlotSpecifications
PIGO | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
class O, DKFZp434M222, EC 2.-, FLJ00135, GPI ethanolamine phosphate transferase 3, MGC20536, MGC3079, phosphatidylinositol glycan anchor biosynthesis, class O, Phosphatidylinositol-glycan biosynthesis class O protein, PIG-O, RP11-182N22.4 | |
A synthetic peptide corresponding to a sequence within amino acids 950-1050 of human PIGO (NP_116023.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 50% glycerol, pH7.3 | |
84720 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title