Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGP Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306346IR
Description
PIGP Polyclonal specifically detects PIGP in Human samples. It is validated for Western Blot.Specifications
PIGP | |
Polyclonal | |
Western Blot | |
class P, DCRCDown syndrome critical region protein C, Down syndrome critical region protein 5, DSCR5DCRC-S, DSCRC, DSRC, EC 2.4.1.198, phosphatidylinositol glycan anchor biosynthesis, class P, Phosphatidylinositol-glycan biosynthesis class P protein, phosphatidylinositol-n-acetylglucosaminyltranferase subunit, PIG-P | |
Produced in rabbits immunized with E. coli-derived Human PIGP fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
51227 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction