Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIGP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555464
Description
Immunogen sequence: NMMSTSPLDS IHTITDNYAK NQQQKKYQEE AIPALRDISI SEVNQMFFLA AKELYTK Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 86%.
Part of the complex catalyzing the transfer of N- acetylglucosamine from UDP-N-acetylglucosamine to phosphatidylinositol, the first step of GPI biosynthesis.
Specifications
PIGP | |
Polyclonal | |
Unconjugated | |
PIGP | |
DCRC; DCRC-S; Down syndrome critical region gene 5; Down syndrome critical region gene c; Down syndrome critical region homolog 5; Down syndrome critical region protein 5; Down syndrome critical region protein 5 homolog; Down syndrome critical region protein C; Dscr5; DSCRC; DSRC; NPD010; phosphatidylinositol glycan anchor biosynthesis class P; phosphatidylinositol glycan anchor biosynthesis, class P; phosphatidylinositol glycan, class P; phosphatidylinositol N-acetylglucosaminyltransferase subunit P; Phosphatidylinositol-glycan biosynthesis class P protein; phosphatidylinositol-n-acetylglucosaminyltranferase subunit; PIGP; PIG-P | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51227 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P57054 | |
PIGP | |
Recombinant protein corresponding to Human PIGP. Recombinant protein control fragment (Product #RP-90557). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction