Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGP Polyclonal Antibody

PIGP Polyclonal Antibody

Supplier:  Thermo Scientific PA5104008

Catalog No. PIPA5104008


Only null left
Add to Cart

Description

Description

PIGP Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PIGP
Polyclonal
Unconjugated
PIGP
DCRC; DCRC-S; Down syndrome critical region gene 5; Down syndrome critical region gene c; Down syndrome critical region homolog 5; Down syndrome critical region protein 5; Down syndrome critical region protein 5 homolog; Down syndrome critical region protein C; Dscr5; DSCRC; DSRC; NPD010; phosphatidylinositol glycan anchor biosynthesis class P; phosphatidylinositol glycan anchor biosynthesis, class P; phosphatidylinositol glycan, class P; phosphatidylinositol N-acetylglucosaminyltransferase subunit P; Phosphatidylinositol-glycan biosynthesis class P protein; phosphatidylinositol-n-acetylglucosaminyltranferase subunit; PIGP; PIG-P
Rabbit
Affinity Chromatography
RUO
51227, 56176
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P57054, Q9JHG1
PIGP
A synthesized peptide derived from human PIGP(Accession P57054), corresponding to amino acid residues I92-E142.
100 μL
Primary
Human, Mouse, Monkey
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.