Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIGP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA548800

Catalog No. PA548800


Description

Description

Part of the complex catalyzing the transfer of N- acetylglucosamine from UDP-N-acetylglucosamine to phosphatidylinositol, the first step of GPI biosynthesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PIGP
Polyclonal
PBS with 0.09% sodium azide
P57054
PIGP
KLH conjugated synthetic peptide between 2-30 amino acids from the N-terminal region of human PIGP.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
PIGP
Phosphatidylinositol N-acetylglucosaminyltransferase subunit P, Down syndrome critical region protein 5, Down syndrome critical region protein C, Phosphatidylinositol-glycan biosynthesis class P protein, PIG-P, PIGP, DCRC, DSCR5, DSCRC
Rabbit
Antigen affinity chromatography
RUO
51227
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.