Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGQ Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | PIGQ |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PIGQ Polyclonal specifically detects PIGQ in Human samples. It is validated for Western Blot.Specifications
| PIGQ | |
| Polyclonal | |
| Rabbit | |
| Q9BRB3 | |
| 9091 | |
| Synthetic peptides corresponding to PIGQ(phosphatidylinositol glycan anchor biosynthesis, class Q) The peptide sequence was selected from the N terminal of PIGQ. Peptide sequence VLHFPFIPIQVKQLLAQVRQASQVGVAVLGTWCHCRQEPEESLGRFLESL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| c407A10.1, c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)), class Q, EC 2.4.1.198, hGPI1, MGC12693, N-acetylglucosaminyl transferase component Gpi1, N-acetylglucosamyl transferase component GPI1, phosphatidylinositol glycan anchor biosynthesis, class Q, phosphatidylinositol N-acetylglucosaminyltransferase subunit Q, Phosphatidylinositol-glycan biosynthesis class Q protein, PIG-Q | |
| PIGQ | |
| IgG | |
| 84 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title