Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGQ Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | PIGQ |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PIGQ Polyclonal specifically detects PIGQ in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PIGQ | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
9091 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:YHTLGLEVIGSEQMWGWPARLAALHHWHCLPWDPLPTCCGHHGGEHSNPRCPEHCPMPTLHTQVQRVRPPQQPQVEGWSPWGLPSGSALAV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
c407A10.1, c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)), class Q, EC 2.4.1.198, hGPI1, MGC12693, N-acetylglucosaminyl transferase component Gpi1, N-acetylglucosamyl transferase component GPI1, phosphatidylinositol glycan anchor biosynthesis, class Q, phosphatidylinositol N-acetylglucosaminyltransferase subunit Q, Phosphatidylinositol-glycan biosynthesis class Q protein, PIG-Q | |
PIGQ | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title