Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGQ Monoclonal Antibody (2B7)
GREENER_CHOICE

Catalog No. PIMA524423
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524423 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524423 Supplier Invitrogen™ Supplier No. MA524423
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: HCPMPTLCTQ VQRVRPPQQP QVEGWSPWGL PSGSALAVGV EGPCQDEPPS PRHPLAPSAE QHPASGGLKQ SLTPVPSGPG PSLPEPHGVY LRMFPGEV.

This gene is involved in the first step in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This gene encodes a N-acetylglucosaminyl transferase component that is part of the complex that catalyzes transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc to phosphatidylinositol (PI).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGQ
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PIGQ
Gene Accession No. Q9BRB3
Gene Alias c407A10.1; c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)); glycosylphosphatidylinositol 1 homolog; GPI1; Gpi1h; Gpi1p; Gpih; Mgpi1; MGpi1p; N-acetylglucosaminyl transferase component Gpi1; N-acetylglucosamyl transferase component GPI1; phosphatidylinositol glycan anchor biosynthesis class Q; phosphatidylinositol glycan anchor biosynthesis, class Q; phosphatidylinositol glycan, class Q; phosphatidylinositol N-acetylglucosaminyltransferase subunit Q; Phosphatidylinositol-glycan biosynthesis class Q protein; Pigq; PIG-Q
Gene Symbols PIGQ
Host Species Mouse
Immunogen PIGQ (NP_683721.1, 661 a.a. ~ 758 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9091
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.