Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIGQ Polyclonal Antibody
PIGQ Polyclonal Antibody
Supplier: Thermo Scientific PA558947
Description
PIGQ Polyclonal Antibody for ICC/IF, IHC

Specifications
PIGQ | |
Polyclonal | |
Unconjugated | |
PIGQ | |
c407A10.1; c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)); glycosylphosphatidylinositol 1 homolog; GPI1; Gpi1h; Gpi1p; Gpih; Mgpi1; MGpi1p; N-acetylglucosaminyl transferase component Gpi1; N-acetylglucosamyl transferase component GPI1; phosphatidylinositol glycan anchor biosynthesis class Q; phosphatidylinositol glycan anchor biosynthesis, class Q; phosphatidylinositol glycan, class Q; phosphatidylinositol N-acetylglucosaminyltransferase subunit Q; Phosphatidylinositol-glycan biosynthesis class Q protein; Pigq; PIG-Q | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9091 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRB3 | |
PIGQ | |
Recombinant protein corresponding to Human PIGQ. Recombinant protein control fragment (Product #RP-95550). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction