Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGQ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558656
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558656 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558656 Supplier Invitrogen™ Supplier No. PA558656
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDQVMLIFYD QRQVLLSQLH LPTVLPDRQA GATTASTGGL AAVFDTVARS EVLFRSDRFD E Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.

This gene is involved in the first step in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This gene encodes a N-acetylglucosaminyl transferase component that is part of the complex that catalyzes transfer of N-acetylglucosamine (GlcNAc) from UDP-GlcNAc to phosphatidylinositol (PI).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGQ
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIGQ
Gene Accession No. Q9BRB3
Gene Alias c407A10.1; c407A10.1 (GPI1 (N-acetylglucosaminyl transferase component)); glycosylphosphatidylinositol 1 homolog; GPI1; Gpi1h; Gpi1p; Gpih; Mgpi1; MGpi1p; N-acetylglucosaminyl transferase component Gpi1; N-acetylglucosamyl transferase component GPI1; phosphatidylinositol glycan anchor biosynthesis class Q; phosphatidylinositol glycan anchor biosynthesis, class Q; phosphatidylinositol glycan, class Q; phosphatidylinositol N-acetylglucosaminyltransferase subunit Q; Phosphatidylinositol-glycan biosynthesis class Q protein; Pigq; PIG-Q
Gene Symbols PIGQ
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIGQ. Recombinant protein control fragment (Product #RP-95549).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.