Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
pIgR Antibody (40), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290428MFV610
Description
pIgR Monoclonal antibody specifically detects pIgR in Mouse samples. It is validated for ELISA, ELISASpecifications
pIgR | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
40 | |
ELISA, Sandwich ELISA Capture | |
FLJ22667, hepatocellular carcinoma associated protein TB6, Hepatocellular carcinoma-associated protein TB6, MGC125361, MGC125362, PIgR, Poly-Ig receptor, polymeric immunoglobulin receptor | |
Rabbit immunized with purified, recombinant Mouse pIgR (Accession#: NP_035212.2, Met 1-Lys 645) | |
0.1 mL | |
Immunology | |
5284 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction