Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGS Monoclonal Antibody (3F3)
GREENER_CHOICE

Catalog No. PIMA524508
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524508 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524508 Supplier Invitrogen™ Supplier No. MA524508
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: GKISNIVIKD DVASEVYKAV AAVQKSAEEL ASGHLASAFV ASQEAVTSSE LAFFDPSLLH LLYFPDDQK.

Component of the GPI transamidase complex. Essential for transfer of GPI to proteins, particularly for formation of carbonyl intermediates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGS
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F3
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PIGS
Gene Accession No. Q6PD26, Q96S52
Gene Alias BC058979; Gm393; Gm689; GPI transamidase component PIG-S; GPI transamidase subunit; phosphatidylinositol glycan anchor biosynthesis class S; phosphatidylinositol glycan anchor biosynthesis, class S; phosphatidylinositol glycan, class S; phosphatidylinositol glycan, class S homolog; phosphatidylinositol-glycan biosynthesis class S protein; PIGS; UNQ1873/PRO4316
Gene Symbols PIGS
Host Species Mouse
Immunogen PIGS (NP_149975.1, 450 a.a. ~ 518 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 276846, 94005
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.