Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553739
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553739 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553739 Supplier Invitrogen™ Supplier No. PA553739
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVLPVRVEVD MVRVMEVFLA QLRLLFGIAQ PQLPPKCLLS GPTSEGLMTW ELDRLLWARS VENLATATTT LTSLAQLLGK ISNIVIKDDV ASEVYKAVAA VQKSAEELAS GHLASAFVAS QEAVTSSELA FFDPSLLHLL YFPDDQ Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Component of the GPI transamidase complex. Essential for transfer of GPI to proteins, particularly for formation of carbonyl intermediates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGS
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIGS
Gene Accession No. Q96S52
Gene Alias BC058979; Gm393; Gm689; GPI transamidase component PIG-S; GPI transamidase subunit; phosphatidylinositol glycan anchor biosynthesis class S; phosphatidylinositol glycan anchor biosynthesis, class S; phosphatidylinositol glycan, class S; phosphatidylinositol glycan, class S homolog; phosphatidylinositol-glycan biosynthesis class S protein; PIGS; UNQ1873/PRO4316
Gene Symbols PIGS
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIGS. Recombinant protein control fragment (Product #RP-90923).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94005
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.