Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIGT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542992
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542992 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542992 Supplier Invitrogen™ Supplier No. PA542992
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LPANSVTKVS IQFERALLKW TEYTPDPNHG FYVSPSVLSA LVPSMVAAKP Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

PIGT is a protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is an essential component of the multisubunit enzyme, GPI transamidase. GPI transamidase mediates GPI anchoring in the endoplasmic reticulum, by catalyzing the transfer of fully assembled GPI units to proteins. This gene encodes a protein that is involved in glycosylphosphatidylinositol (GPI)-anchor biosynthesis. The GPI-anchor is a glycolipid found on many blood cells and serves to anchor proteins to the cell surface. This protein is an essential component of the multisubunit enzyme, GPI transamidase. GPI transamidase mediates GPI anchoring in the endoplasmic reticulum, by catalyzing the transfer of fully assembled GPI units to proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIGT
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PIGT
Gene Accession No. Q969N2
Gene Alias 2510012P17Rik; 4930534E15Rik; CGI-06; FLJ41596; GPI transamidase component PIG-T; GPI transamidase subunit; MCAHS3; MGC8909; NDAP; Ndap7; Neuronal development-associated protein 7; neurotrophin-regulated neuronal development-associated protein; phosphatidylinositol glycan anchor biosynthesis class T; phosphatidylinositol glycan anchor biosynthesis, class T; phosphatidylinositol glycan, class T; phosphatidylinositol-glycan biosynthesis class T protein; PIGT; PNH2; PSEC0163; si:ch211-153a8.2; UNQ716/PRO1379
Gene Symbols PIGT
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PIGT (aa 434-483).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51604
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.